MagLab™ Lab Storage Series
Supplier: Heathrow Scientific
HS23500 HS23502 HS23503 HS23504
89234-768CS 3261.43 CAD
89234-768 89234-772 89234-774 89234-776
MagLab™ Lab Storage Series
Racks Tube Racks
Easily organize common lab products off your benchtop with the MagLab storage system.
  • Made of robust ABS plastic in bright translucent colors
  • Secure direct to metal with built-in powerful magnets or non-metal surfaces with included self-adhesive mounting plate
Create lab storage space-off your benchtop.
*Kimwipes is a registered trademark of Kimberly-Clark Corp.
Each rack is made of robust ABS plastic in a bright translucent color and features four powerful magnets for a secure hold onto most metal surfaces. MagRack™ holds four 50 ml tubes andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable