Stopcock, Inlet Feed Tube, Ace Glass Incorporated
Supplier: Ace Glass
13295-20 13295-08 13295-10
89077-634EA 154.08 CAD
89077-634 89083-150 89083-152
Stopcock, Inlet Feed Tube, Ace Glass Incorporated
Stopcocks
This stopcock is used in Heidolph 4000 Series rotary evaporator condensers and other popular brand rotary evaporator condensers as an inlet feed tube to allow continuous feed of volumes exceeding capacity of evaporating flask
Joint size is [ST] 19/38. Fabricated from borosilicate glass.