m-ColiBlue24® Media, Hach
Supplier: Hach
2608450 2608420 2608442
CA11016-726PK 527.67 CAD
CA11016-726 CA11016-728 CA11016-730
m-ColiBlue24® Media, Hach
Media for Microbiology Prepared Media for Microbiology
In 24 hours, Hach's m-ColiBlue24® Broth give you simultaneous results for total coliforms and E. coli.
  • Simultaneous results
  • Ready-to-use
  • Evaluate total coliform and E. coli
Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media
Use m-ColiBlue24® Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.
m-ColiBlue24 should only be used with pads manufactured by the following companies/brands: Pall Corporation (e.g., Sterile Petri Dishes with Absorbant Pads), Private Label/OEM products manufactured by Pall Gelman, Sartorius (cellulose and glass fiber pads), and Sartorius BioSart products with the integrated Petri dish. No other pad is compatible with the product, and the use of any pad other than those recommended may result in unreliable, inaccurate data.
With ready-to-use m-ColiBlue24® Broth, you can simultaneously detect total coliforms and E. coli—in just 24 hours, easily read and interpret results, save money by eliminating special equipment and UV lamps, minimize background growth of non-coliform bacteria, reduce false positives and false negatives, plus get superior sensitivity down to 1 CFU/100 mL, and conduct emergency testing and routine monitoring.
Order Now
SPECIFICATIONS
Environmentally Preferable