BART Test for Iron-Related Bacteria, Hach
Supplier: Hach
2432327 2432309 2432309
CA89000-176PK 597.32 CAD
97026-410 97026-620 CA89000-176
BART Test for Iron-Related Bacteria, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence of process disruptors, such as iron-reducing bacteria (IRB) that could be eating your pipes.
  • Identify the presence of process disruptors
  • Monitor the population size of bacteria
  • Easy to use
An important agent involved in biofouling, IRB can cause corrosion, clogging, fouling, and hygiene risk.
Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
Easy to use, requiring no elaborate or costly equipment and no specialized training.
Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment.
Certifications: UL Listed.
Accessories information: Accessory kit includes plug and case.