Meander Fusion Chamber, BTX™
Supplier: BTX
45-0107
58017-992PK 3501.06 CAD
58017-992
Meander Fusion Chamber, BTX™
Electroporation Systems Electroporators
The BTX™ Meander Fusion Chamber is a novel microslide design which is specifically used for electro cell fusion
The Meander Fusion Chamber generates a divergent field and is used for fusion of mammalian cells, plant, yeast, fungi, and microorganisms. The specialty electrode is constructed of a conductive metal alloy. It has two primary bars that are connected by many tiny fingerlike projections. These projections are spaced 0.2mm apart. The electrode is mounted on a glass slide. It is designed for direct viewing of dimer formation during alignment while under a microscope.
Ordering information: The Meander Fusion Chamber is supplied with 0.2mm gap chamber in a pack of 4. Connection cables are required.