Spectra/Por Regenerated Cellulose Dialysis Discs and Sheets, Spectrum Laboratories
Supplier: Spectrum Laboratories
132476 132478 132488 132677 132723 132480 132482 132486 132686
25213-101PK 551.4 CAD
25213-101 25213-102 25213-103 25213-180 25213-188 25224-303 25224-347 25224-369 97059-124 25213-107 25213-147 25213-149 25213-178 25213-186 25213-227 25224-325
Spectra/Por Regenerated Cellulose Dialysis Discs and Sheets, Spectrum Laboratories
Tubing Dialysis Tubing
Pre-cut single layer membrane discs and sheets for a variety of dialysis equipment.
Available in a variety of dimensions to meet your application needs. Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes.