Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
12770-12 12770-14 12770-16 12770-18 12770-24 12770-26 12770-27 12770-28 12770-30 12770-31 12770-32 12770-36 12770-38 12770-52 12770-54 12770-59
89077-502EA 168.44 CAD
89077-502 89077-504 89077-506 89077-508 89077-510 89077-512 89077-514 89077-516 89077-518 89077-520 89077-522 89077-524 89077-526 89077-528 89077-530 89077-532
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Fittings and Connectors
These all-PTFE fittings have a male NPT (MNPT) thread at one end and either tube compression, a hose barb, or MNPT thread at the other end.