Syringe Gas Tight, Ace Glass Incorporated
Supplier: Ace Glass
5931-01 5931-02 5931-03 5931-04 5931-06
89053-698EA 131.91 CAD
89053-698 89053-700 89053-702 89053-704 89053-706
Syringe Gas Tight, Ace Glass Incorporated
Syringes Syringes for Chromatography
This syringe is designed for the highest performance in applications such as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions
PTFE coated plungers with precision PTFE tips for leak-tight seal. Accuracy and reproducibility of ±1%. Has a fixed needle.
Ordering information: Several sizes of this syringe are available with Luer-Lok tip and Luer-Lok. Contact your Avantor representative for more information.