Connectors, with PTFE Plug Valve, Ace Glass Incorporated
Supplier: Ace Glass
12611-04 12611-08 12611-21 12611-23
71000-282CS 950.39 CAD
71000-282 71000-284 71000-286 71000-288
Connectors, with PTFE Plug Valve, Ace Glass Incorporated
Valves Non Return Valves
Polypropylene connectors, two-way or three-way, with PTFE valve and serrated tubulations
Takes 6.4 mm to 9.5 mm I.D. tubing.