PTFE Stopcock Valve, Two Way, Ace Glass Incorporated
Supplier: Ace Glass
5839-04 5839-08 5839-14 5839-42 5839-47 5839-55 5839-10 5839-62 5839-68 5839-72 5839-78
89052-608EA 147.64 CAD
89052-608 89052-610 89052-614 89052-616 89052-618 89052-620 89052-612 10099-046 10099-048 10099-050 10099-052 10099-054 10099-056 10099-058 10099-060 10099-062 10099-064
PTFE Stopcock Valve, Two Way, Ace Glass Incorporated
Valves Non Return Valves
This PTFE valve is used with NPT fittings.
  • Various ends available
  • Stopcocks and compression ferrules made of virgin PTFE
  • Nuts and ferrule-gripper made of PVDF
Various ends available, including male and female NPT, luer, ¼" tube and ¼"-28 UNF. IPS PTFE Stopcocks are made of virgin PTFE. Compression ferrules are also made of virgin PTFE. Nuts and ferrule-gripper are made of PVDF.
Order Now
SPECIFICATIONS
Environmentally Preferable