Flask, Reaction, Flat Flange, O-Ring Groove, Flat Bottom, Ace Glass Incorporated
Supplier: Ace Glass
6511-53 6511-56 6511-58 6511-60
89055-074EA 663.65 CAD
89055-074 89055-076 89055-078 89055-080
Flask, Reaction, Flat Flange, O-Ring Groove, Flat Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Cylindrical heavy-wall reaction flask, with flat bottom rounded inside wall, and flat flange
Flange has an o-ring groove for use with CAPFE® (PTFE encapsulated silicone rubber) o-ring instead of a gasket. Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Fabricated from borosilicate glass.
Ordering information: Flask is supplied with one CAPFE o-ring, 89061-176 for 1000mL size, 89061-178 for 2000, 3000 and 4000mL sizes.