Flask, Reaction, Flat Flange, Flat Bottom, Ace Glass Incorporated
Supplier: Ace Glass
6511-24 6511-27 6511-29 6511-31
89055-058EA 554.77 CAD
89055-058 89055-060 89055-062 89055-064
Flask, Reaction, Flat Flange, Flat Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Cylindrical heavy wall reaction flask, with flat bottom rounded into side wall, and flat, ground flange top
Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Fabricated from borosilicate glass.