Needles, Special Length, Ace Glass Incorporated
Supplier: Ace Glass
13684-07 13684-11 13684-15 13684-19
89078-061CS 2083.33 CAD
89078-061 89078-063 89078-065 89078-067
Needles, Special Length, Ace Glass Incorporated
Syringe Needles
Special length 304 stainless steel needles with a deflected septum point and standard female hub
The deflected septum point is equivalent to B-D Huber or Hamilton Style 1 and 2.
Packaging: Packed 6 per package or a case of 12.
Order Now
SPECIFICATIONS
Environmentally Preferable