Clamp, Three-Prong, Double Screws, Ace Glass Incorporated
Supplier: Ace Glass
11065-17 11069-18 11067-14 11065-16
89083-246EA 199.66 CAD
89083-246 89083-248 89083-250 89083-252
Clamp, Three-Prong, Double Screws, Ace Glass Incorporated
Clamps General Purpose Clamps
Three-prong grip clamp with double screws
Because both jaws are adjustable, with double screws that operate independently, large or small objects can be held easily. Comes with an extra set of fiberglass prong covers for temperatures above 100°C.