Chromatography Column, 1:5 PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
5888-10 5888-15 5888-20 5888-25 5888-30 5888-35 5888-37 5888-41 5888-45 5888-50 5888-39 5888-42 5888-55
89053-486EA 264.72 CAD
89053-486 89053-488 89053-490 89053-492 89053-494 89053-496 89053-498 89053-502 89053-506 89053-508 89053-518 89053-520 89053-524
Chromatography Column, 1:5 PTFE Plug, Ace Glass Incorporated
Chromatography Columns
This chromatography column has a 2 mm bore 1:5 taper solid PTFE stopcock plug at bottom.
Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable