VWR® General Purpose Digital Laboratory Incubators, 10.8 cubic feet
Supplier: VWR International
1556 1556-2
97058-224EA 18436.63 CAD
97058-224 97058-226
VWR® General Purpose Digital Laboratory Incubators, 10.8 cubic feet
Incubators Standard Incubators
General-purpose incubators deliver a degree of temperature uniformity usually found only in more expensive, application specific models.
  • Features include: Microprocessor control, independent over temperature, thermostat, unique warm air jacket design, interior outlet, heated outer door, viewing door, stainless steel, interior, access port
This high level of performance is due to a precise microprocessor controller and a unique warm air jacket design
This design does not require blowers or fans within the incubator chamber where samples are susceptible to cross-contamination and the drying effects of convection. Five strategically placed heating elements, including a heated outer door, ensure uniform heating even when the incubator chamber is fully loaded.
Triple wall construction allows heated air to circulate within an open plenum between the inner chamber and exterior walls.