Orbital-Genie™ Large Capacity Orbital Shaker, Scientific Industries
Supplier: Scientific Industries
SI-1500 SI-1600 SI-1700 SI-1610 SI-1611 SI-1612 SI-1613 SI-1614 SI-1615 SI-1617 SI-1510 SI-1618 SI-1616 SI-1616
97060-146EA 7329.71 CAD
97060-146 97060-150 97060-154 97060-158 97060-160 97060-162 97060-164 97060-166 97060-168 97060-170 97060-172 89401-426 89426-878 490010-030
Orbital-Genie™ Large Capacity Orbital Shaker, Scientific Industries
Shakers Orbital Shakers
The Orbital-Genie™ Large Capacity Orbital Shaker is a powerful large capacity (460×460 mm platform) orbital shaker designed to hold any type of container.
  • Holds any type of container
  • Fully programmable
  • Variable speed
  • Three platform configurations
Fully programmable shaker features variable speed including orbit reverse, timer from 1 minute to 99 hours with alarm, programmable acceleration and deceleration rates, programmable ON/OFF times for interval shaking. Operator can create any mixing or shaking action from smooth vortex to sloshing aeration. Available in three platform configurations: Universal Ratcheting Clamps - provide the flexibility for using a mix of different size and shape vessels on a single platform. Adhering Mat - provides a flat platform with adhering rubber mat perfect for use with open containers, especially large containers. Standard Fask Clamps - is equipped with a platform for accepting various size flasks or beakers. (Clamps sold separately). All units can be used in a cold room or incubator and CO₂ environments.
Certifications: CE marked.
Ordering information: Two ratcheting and one non-ratcheting cushioned clamps included with Ratcheting Clamps model.