Joint/Flask Adapters Without Combi Clip, Ace Glass Incorporated
Supplier: Ace Glass
3954-11 3954-06 3954-08 3954-04
89083-050EA 467.3 CAD
89083-050 89083-100 89083-102 89083-098
Joint/Flask Adapters Without Combi Clip, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a replacement receiving flask adapter for Buchi rotary evaporators.
  • For Buchi rotary evaporators
  • Without combi clip
  • Fabricated from borosilicate glass
Joint to flask adapter joins the steam tube to the receiving flask. Fabricated from borosilicate glass.