Rotator Genie™ Rotator, Scientific Industries
Supplier: Scientific Industries
SI-2100 SI-2200 SI-2110 SI-2210
89202-302EA 1989.01 CAD
89202-302 89202-306 89202-310 89202-312
Rotator Genie™ Rotator, Scientific Industries
Shakers Rotating/Rocking Shakers
This Rotator is a compact but rugged lab rotator available in two interchangeable platform configurations providing the versatility to hold virtually any kind of container.
  • Rugged construction
  • Interchangeable configurations
  • Holds any kind of container
Choose from a Standard Platform with Retaining Bands model (no additional accessories necessary), or a Stainless Steel Magnetic Platform model with clip plate system (clip plates sold separately). Uniquely designed for a wide selection of motions from gentle rolling to forceful end over end mixing. Ideal for Cold Rooms and Incubators. (Will easily fit in most incubators).
Certifications: CE marked.
Ordering information: Standard Platform with Retaining Bands model does not require additional accessories. Magnetic Platform model requires clip plates sold separately. Platforms are interchangeable.