Cap, GL, with Hole, Ace Glass Incorporated
Supplier: Ace Glass
7621-15 7621-25 7621-04 7621-08 7621-22
89059-778EA 10.95 CAD
89059-778 89059-776 89059-774 89059-782 89059-780
Cap, GL, with Hole, Ace Glass Incorporated
Closures Bottle Closures
This open top red polybutylene teraphthalate (PBT) cap is for use with GL threads
When used with a hose connection and appropriate sealing ring, this cap will accommodate tubing of the same approximate diameter. Temperature range is –45ºC to 180ºC.