Vigreux Distilling Column, Ace Glass
Supplier: Ace Glass
6578-30 6578-12 9345-09 9345-10 6578-04 6578-16 6578-24 6578-08 6578-34 6578-10 9345-13 6578-20 6578-22 6578-06 9345-08 9345-11 6578-14 6578-32
89055-664EA 485.42 CAD
89055-664 76214-398 89074-422 89074-420 89055-662 76214-402 89055-660 89074-424 76214-400 89074-418 89074-416 89055-658 89055-668 89055-656 89055-666 76214-406 76214-404 76214-408
Vigreux Distilling Column, Ace Glass
Distillation Components Distillation Columns
The column length in millimeters (listed below) refers to the effective length of the column as measured from the lowest to the highest indent of the column
Outer [ST] joint at top, ground [ST] inner joint at bottom. Full-sized joints (only) are tooled with reinforcement rings for added strength and stability. Fabricated from borosilicate glass.