Chromatography Column, with Fritted Disc, Ace Glass Incorporated
Supplier: Ace Glass
5885-06 5885-12 5885-18 5885-24
89053-478EA 137.84 CAD
89053-478 89053-480 89053-482 89053-484
Chromatography Column, with Fritted Disc, Ace Glass Incorporated
Chromatography Columns
Plain, open top, chromatography column, with a Porosity B (70 to 100 µm) fritted disc sealed in.
Fabricated from borosilicate glass.