Trap, Rotary Evaporator, Anti-Splash, Ace Glass Incorporated
Supplier: Ace Glass
6705-06 6705-10 6705-14 6705-22 6705-26 6705-12
89055-874EA 377.11 CAD
89055-874 89055-876 89055-878 89055-880 89055-882 89085-004
Trap, Rotary Evaporator, Anti-Splash, Ace Glass Incorporated
Vacuum Traps
This rotary evaporator trap is designed to greatly reduce carry-over from foaming or bumping.
Two drain holes, close to base of the inner tube, allow solvent to drain back into evaporator flask. Fabricated from borosilicate glass.