Addition Funnel, Cylindrical, Ace Glass Incorporated
Supplier: Ace Glass
5822-05 5822-15 5822-20 5822-10
89052-422EA 221.27 CAD
89052-422 89052-426 89052-428 89052-432
Addition Funnel, Cylindrical, Ace Glass Incorporated
Funnels Dropping Funnels
This funnel is used for charging columns
The capacity is approximately five times the largest column volume. Size listed is Ace-Thred designation which refers to I.D. of tubing above thread and corresponds to the column diameter. Fabricated from borosilicate glass.