Addition Funnel, Conical Shape, Ace Glass Incorporated
Supplier: Ace Glass
5822-45 5822-40 5822-50 5822-55
89052-424EA 233 CAD
89052-424 89052-430 89052-434 89052-436
Addition Funnel, Conical Shape, Ace Glass Incorporated
Funnels Dropping Funnels
This funnel is used for charging columns
The capacity is approximately five times the largest column volume. Size listed is Ace-Thred designation which refers to I.D. of tubing above thread and corresponds to the column diameter. Fabricated from borosilicate glass.