Adapter, Feed Tube, Ace Glass Incorporated
Supplier: Ace Glass
5827-20 5827-24 5827-28 5827-30
89052-462EA 173.41 CAD
89052-462 89052-464 89052-466 89052-468
Adapter, Feed Tube, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is threaded at both ends and on side joint.
  • Top and side nylon bushings and FETFE o-rings included
  • One silicone rubber septa also included
  • Fabricated from borosilicate glass
The #7 Ace-Thred joints at top and side are each supplied with a nylon bushing and FETFE o-ring. One silicone rubber septa is also included. Size listed also refers to the I.D. of the tubing above the lower thread, and this corresponds to the column diameter.