Adapter, 4mm Bottom Drip, Ace Glass Incorporated
Supplier: Ace Glass
5834-05 5834-10 5834-15 5834-20
89052-070EA 112.05 CAD
89052-070 89052-072 89052-500 89055-804
Adapter, 4mm Bottom Drip, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This drip tip adapter is used for chemical type chromatography.
  • Top Ace-Thred
  • Bottom drip tip
  • Fabricated from borosilicate glass
The I.D. of drip is approximately 4mm. Ace-Thred size listed also refers to the I.D. of the tubing below thread, and corresponds to the column diameter.