HyClone™ FetalClone™ II serum, U.S. Origin, HyClone products (Cytiva)
Supplier: Cytiva
SH30066.03 SH30066.02 30066.03IH2540 3006603IR25-40 3006603IR30-45 SH30066.02HI SH30066.03 SH30066.03HI
16777-236EA 829.36 CAD
16777-236 CA16777-234 CA76499-992 CA76324-854 CA76534-646 CA76237-658 CA16777-236 CA82013-594
HyClone™ FetalClone™ II serum, U.S. Origin, HyClone products (Cytiva)
Sera (Serum)
HyClone™ FetalClone® II is a foetal bovine serum alternative that contains additional growth factors and supplements in the formulation. It has been developed specifically for growth of CHO cells and derivatives.
  • IgG levels comparable with those found in FBS
  • Improved cell growth performance relative to FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0,1 µm) pore size rated filters. Each lot undergoes extensive quality performance and analytical testing.