Adapter, Bottom Drip, 1:5 PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
5835-07 5835-11 5835-17 5835-21
89052-504EA 382.16 CAD
89052-504 89052-506 89052-508 89052-510
Adapter, Bottom Drip, 1:5 PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Drip tip adapter for chemical type chromatography, with 1:5 taper PTFE stopcock at bottom for controlling flow.
  • 1:5 taper PTFE stopcock at bottom
  • Top Ace-Thred
  • Fabricated from borosilicate glass
The Ace-Thred size also refers to the I.D. of the tubing below the thread, and corresponds to the column diameter.