Bottom Outlet Valve/Adapter for Big Columns, Ace Glass Incorporated
Supplier: Ace Glass
5835-32 5835-34
89052-512EA 446.62 CAD
89052-512 89052-514
Bottom Outlet Valve/Adapter for Big Columns, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This bottom outlet valve/adapter is used with big chromatography columns
Two versions are available: one has a 0 – 8 mm threaded stopcock for controlling flow; The second has a straight stem without stopcock. Both take 3/8" Swagelok fittings, and they can be secured to a bottom adapter via 89059-216 front-seal bushing.