UltraJaws 3-Prong Extension Clamp, Ace Glass Incorporated
Supplier: Ace Glass
11064-13 11064-11 11064-09
89083-198EA 188.07 CAD
89083-198 89083-200 89083-202
UltraJaws 3-Prong Extension Clamp, Ace Glass Incorporated
Clamps General Purpose Clamps
These multi-purpose precision clamps are a new, innovative design, with closed-yoke construction that minimizes corrosion and contamination of internal components
This unique design enables very secure gripping and positioning. All clamps are three-prong with dual adjustment screws. Clamps are supplied with non-slip vinylized jaws and a set of extra fiberglass prong covers for temperatures above 100ºC.