Distillation Column, Vigreux, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
9345-32 9345-35
89074-426EA 225.6 CAD
89074-426 89074-428
Distillation Column, Vigreux, Microscale, Ace Glass Incorporated
Distillation Components Distillation Columns
Vigreux columm for vacuum distillation of high boiling materials
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground [ST] 14/10 outer joint. Bottom is a [ST] 14/10 inner joint, and indents. Length stated is length of indents. Fabricated from borosilicate glass.
Ordering information: Supplied with open-top phenolic cap and FETFE o-ring.