HyClone™ SFM4Insect Media with L-Glutamine, HyClone products (Cytiva)
Supplier: Cytiva
HyClone™ SFM4 insect is a versatile cell culture medium developed through Metabolic Pathway Design to support the growth of multiple insect cell lines and production of a variety of recombinant proteins without compromising performance. HyClone SFM4 insect provides superior growth of many key insect cell lines, including Sf9, Sf21, High Five (T.ni), and D.mel.
- Supports multiple insect cell lines
- Free of animal derived components
- Protein-free
- High performance growth in multiple culture systems, including high density bioreactors
- Full component traceability
- Manufactured in cGMP and ISO compliant facility
Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).