HyClone™ CDM4HEK293, HyClone products (Cytiva)
Supplier: Cytiva
SH30859.01
CA95026-630EA 140.13 CAD
CA95026-630 CA76462-536
HyClone™ CDM4HEK293, HyClone products (Cytiva)
Cell Culture Media
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
  • Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.