Erlenmeyer Flask, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
9471-42 9471-44
89074-844EA 71.4 CAD
89074-844 89074-846
Erlenmeyer Flask, Microscale, Ace Glass Incorporated
Flasks Erlenmeyer Flasks
Microscale Erlenmeyer flask on which the top joint is a combination cap, 20-400 thread, and joint to accommodate and seal other microscale apparatus.
The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground [ST] 14/10 outer joint.
Ordering information: Supplied with holed cap, PTFE-faced silicone rubber septum, and 89060-994 FETFE o-ring (size –112).
Order Now
SPECIFICATIONS
Environmentally Preferable