Plug Stoppers with Front Seal, Nylon or PTFE, Ace Glass Incorporated
Supplier: Ace Glass
5846-12 5846-14 5846-16 5846-50 5846-51 5846-52 5846-60 5846-04 5846-06 5846-18 5846-22 5846-27 5846-44 5846-46 5846-48 5846-49 5846-45
89052-802EA 76.13 CAD
89052-802 89052-804 89052-806 89052-820 89052-822 89052-824 89052-826 89052-828 89052-830 89052-838 89052-840 89052-842 89052-844 89052-846 89052-848 89052-850 89084-370
Plug Stoppers with Front Seal, Nylon or PTFE, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
Solid plug stoppers are designed for sealing column ends
Plug stoppers feature a "front seal," meaning the O-Ring on the Ace-Thred connection is located at the bottom of the threads rather than the top. This provides a better, more consistent seal. This is critical for pressure applications, and it also prevents viscous materials from migrating up into the threads.
They permit preparation and storage of columns. Each stopper is supplied with one FETFE O-Ring, with the exception of item 89084-370, which is supplied with a CAPFE O-Ring. Plug stoppers are available in Nylon or PTFE, withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable