Pear-Shaped Recovery Flask, Ace Glass Incorporated
Supplier: Ace Glass
9470-26 6892-211 9470-223 6892-212 6892-293 6892-218 6892-213 6892-240 9470-21 6892-222 9470-10 9470-202 9470-225 9470-02 6892-42 6892-32 6892-07 6892-09 6892-14 6892-206 9470-208 6892-18 6892-49 6892-55 6892-237 6892-08 6892-13 6892-12 6892-242 6892-245 6892-208 6892-209 9470-226 6892-205 6892-53 6892-231 6892-216 6892-207 6892-230 6892-40 9470-221 9470-206 9470-04 6892-11 6892-16 6892-249 9470-210 6892-04 6892-204 6892-51 6892-57 9470-43 6892-243 6892-06 9470-227 6892-30 6892-239 9470-27 6892-247 9470-45 9470-25 6892-203 6892-22 6892-214 6892-05 9470-06 6892-15 6892-217 9470-08 9470-204 6892-232 9470-23
89074-788EA 91.01 CAD
89074-788 89074-822 89074-820 89083-862 89074-826 89083-864 89074-824 89083-866 89083-868 89074-828 89056-342 89056-344 89056-300 89056-340 89056-306 89056-308 89056-302 89056-346 89056-304 89056-348 89215-080 89074-796 89215-082 89074-794 89215-084 89074-830 89215-086 89074-798 89056-310 89056-354 89056-312 89056-350 89056-352 89056-318 89056-314 89074-792 89056-316 89074-790 89083-606 89215-074 89074-800 89215-076 89074-804 89215-078 89074-802 89083-602 89074-808 89083-604 89074-806 89056-320 89056-322 89056-328 89056-324 89056-326 89074-810 89074-814 89215-066 89083-872 89056-290 89074-812 89056-292 89074-818 89074-816 89056-332 89056-298 89056-334 89056-294 89056-330 89083-870 89056-296 89056-336 89056-338
Pear-Shaped Recovery Flask, Ace Glass Incorporated
Flasks Round Bottom Flasks
Multipurpose, pear-shaped flasks feature a modified side for easier insertion of spatulas or brushes for removal of solids, or for cleaning
Suitable for use with rotary evaporators. Available in heavy or standard wall. Heavy wall flasks are fabricated with 30% heavier walls than standard-wall flasks. With [ST] outer joint neck. Fabricated from borosilicate glass.