Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Supplier: Scientific Plastics
W39081-150 F164L081-150 W39081-154 F164L081-154 W39081-158 F164L081-158
89209-720EA 402.57 CAD
89209-720 89209-722 89209-724 89209-726 89209-728 89209-730
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Faucets and Sinks Sinks
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
  • Beveled sides help reduce splash
  • Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.