Syringe, Gas Tight, Ace Glass Incorporated
Supplier: Ace Glass
5933-03 5933-05 5933-07 5933-09 5933-11 5933-13
89053-710EA 219.94 CAD
89053-710 89053-712 89053-714 89053-716 89053-718 89053-720
Syringe, Gas Tight, Ace Glass Incorporated
Syringes Syringes for Chromatography
Gas tight syringes with removable needle type (RN) and a 3/4-inch length, bevel point style #2 needle
Used for chromatography and other applications.