Gas Dispersion Tube, Fine Dispersion, Ace Glass Incorporated
Supplier: Ace Glass
9435-09 9435-10 9435-06 9435-25 9435-24 9435-51 9435-21 9435-55 9435-40 9435-38 9435-07 9435-08 9435-39 9435-53 9435-36 9435-22 9435-54 9435-37 9435-52 9435-23
89074-498EA 69.07 CAD
89074-498 89074-510 89074-496 89074-502 89074-500 89083-466 89074-506 89083-476 89083-468 89074-504 89083-458 89074-508 89083-460 89083-470 89083-462 89083-472 89083-464 89083-474 89074-494 89074-492
Gas Dispersion Tube, Fine Dispersion, Ace Glass Incorporated
Gas Tubes
This gas dispersion tube is for fine dispersion
Porous tip is approximately 5mm or 7mm O.D. x 10mm length. The 7mm O.D. tubes fit a #7 Ace-Thred. Fabricated from borosilicate glass.