Round-Bottom Receiving Flasks, [SJ], Ace Glass Incorporated
Supplier: Ace Glass
6902-226 6902-227 6902-228 6902-229 6902-234 6902-235 6902-238 6902-240 6902-241 6902-242 6902-243 6902-244 6902-25 6902-26 6902-27 6902-28 6902-29 6902-34 6902-38 6902-40 6902-41 6902-42 6902-43 6902-44 3996-20 3996-22 3996-24 3996-04 3996-06 3996-08 3996-02
89056-372EA 144.22 CAD
89056-372 89056-374 89056-376 89056-378 89056-380 89056-382 89056-384 89056-386 89056-388 89056-390 89056-392 89056-394 89056-396 89056-398 89056-400 89056-402 89056-404 89056-406 89056-408 89056-410 89056-412 89056-414 89056-416 89056-418 89083-132 89083-134 89083-136 89215-090 89215-092 89215-094 89215-096
Round-Bottom Receiving Flasks, [SJ], Ace Glass Incorporated
Flasks Rotary Evaporator Flasks
Round-bottom, single neck receiving flasks with a short neck and a [SJ] socket joint
Suitable for use with all rotary evaporators. Available with heavy wall (safety coated or uncoated), and with standard walls (uncoated). Heavy wall flasks feature walls that are approximately 30% heavier than standard wall flasks. Fabricated from borosilicate glass