Coupling, Michel-Miller, Ace Glass Incorporated
Supplier: Ace Glass
5805-03 5805-08 5805-15 5805-20 5805-28 5805-32
89052-116EA 267.54 CAD
89052-116 89052-118 89052-120 89052-122 89052-124 89052-126
Coupling, Michel-Miller, Ace Glass Incorporated
Fittings and Connectors
These precision made PTFE couplings have a 316 stainless steel liner for connecting Michel-Miller and other columns when packing.
Designed to make a leak-tight seal without the use of o-rings. Simply tighten until white ring appears indicating a seal has been made, and secure lock nut. No need to tighten further. Male thread is size designation, and matches thread size of columns.