Adjusta-Chrom, Recycling Column, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5819-03 5819-02 5819-12 5819-34 5819-36 5819-13 5819-14 5819-15 5819-26 5819-27 5819-28 5819-44
89052-226EA 2191.13 CAD
89052-226 89052-228 89052-234 89052-252 89052-256 89052-278 89052-282 89052-284 89052-300 89052-302 89052-304 89052-310
Adjusta-Chrom, Recycling Column, Jacketed, Ace Glass Incorporated
Chromatography Columns
Adjustable bed chromatography columns used for recycling, ascending, or descending applications.
Completely jacketed for better temperature uniformity, with a single bushing at each end allowing for quick and easy adjustment of the PTFE plunger. The Porosity C (25 to 50 µm) glass disc gives rapid flow with a minimum of mixing. Pressure tight to 3.515 kg/cm² (50 psi) at room temperature or slightly below. PTFE plunger available with PTFE or O-ring seals. Jacket has two Size D hose connections, for use with ³/₈" int.Ø tubing. Fabricated from borosilicate glass.
Ordering information: Complete item includes jacketed column, plunger tip, appropriate extender(s) with screw cap(s), nylon bushing, PTFE tubing. These items are also available separately. Contact your Avantor representative for more information.