Adapter, Bottom Drip, NPT Thread, with Glass Disc, Ace Glass Incorporated
Supplier: Ace Glass
5838-91 5838-94 5838-96
89052-602EA 201.08 CAD
89052-602 89052-604 89052-606
Adapter, Bottom Drip, NPT Thread, with Glass Disc, Ace Glass Incorporated
Fittings and Connectors
For use as a column end fitting that allows connection of appropriate size Ace-Thred to tubing connectors with NPT threads.
  • Glass disc
  • Fabricated of PTFE
  • Supplied complete with one glass filter disc and one CAPFE o-ring
Adapters are fabricated of PTFE and supplied complete with one 25-50 micro Porosity C glass filter disc and one CAPFE o-ring.
Ordering information: For optional filter disc remover, order 89052-532.