Adapter, End Fitting, NPT to Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5802-04 5802-08 5802-12 5802-14 5802-15 5802-17 5802-19 5802-27 5802-37 5802-40
89052-088EA 184.85 CAD
89052-088 89052-090 89052-092 89052-094 89052-096 89052-098 89052-100 89052-102 89052-104 89052-106
Adapter, End Fitting, NPT to Ace-Thred, Ace Glass Incorporated
Fittings and Connectors
This precision-made PTFE adapter is used for making NPT thread connection to Ace-Threds.
  • Leak-tight seal without o-rings
  • One end seals with Ace-Thred
  • Other end accepts NPT male thread
One end of adapter is designed to make a leak-tight seal with the Ace-Thred without the use of o-rings, while the other end accepts an NPT male thread.