Stopper, Michel-Miller, Ace Glass Incorporated
Supplier: Ace Glass
5803-05 5803-08 5803-11 5803-25
89052-108EA 181.64 CAD
89052-108 89052-110 89052-112 89052-114
Stopper, Michel-Miller, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
These precision made PTFE stoppers are for use at either end of Michel-Miller columns, or on other Ace-Thred connections, when a leak-tight seal without the use of o-rings is desired