Bottom Adapter for Big Columns, Ace Glass Incorporated
Supplier: Ace Glass
5857-30 5857-35 5857-44 5857-46 5857-60 5857-64 5857-67 5857-69 5857-72
89053-002EA 361.54 CAD
89053-002 89053-008 89053-014 89053-016 89053-022 89053-024 89053-026 89053-030 89053-032
Bottom Adapter for Big Columns, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This end-fitting adapter fits large columns with #80 Ace-Thred connections.
  • Recessed
  • Two bottom options
  • Available in UHMWPE or PTFE
  • Available complete or as adapter only
Adapter is recessed to allow the perforated support plate and retaining ring to fit. Bottom outlet options include: a #11 Ace-Thred that connects to a front seal 89059-216 Ace-Thred nylon bushing (with 89061-046 FETFE o-ring) to secure the outlet valve; or a thread that will accept a 3/8-inch NPT fitting. Choose from Ultra High Molecular Weight Polypropylene (UHMWPE) or PTFE. Available as complete item, or adapter only.
Ordering information: Complete item includes appropriate adapter, 89052-914 perforated glass support plate, PTFE (89053-020) or UHMWPE (89053-018) retaining ring, and 89059-216 bottom bushing or connector.