Adapter, Column, Feed Tube, Ace Glass Incorporated
Supplier: Ace Glass
5826-20 5826-24 5826-28 5826-30
89052-452EA 136.72 CAD
89052-452 89052-454 89052-456 89052-458
Adapter, Column, Feed Tube, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is threaded at both ends.
  • Top is #7 Ace-Thred
  • Lower thread
  • Fabricated from borosilicate glass
The top of this adapter has a #7 Ace-Thred that is supplied with a nylon bushing and FETFE o-ring for use with adjustable feed tubes. Size listed also refers to the I.D. of the tubing above the lower thread, and this corresponds to the column diameter.
Ordering information: For extra bushings (includes two FETFE o-rings), order 89051-122. For o-ring only, order 89060-958 (package of 12).