Adapter, Bottom Drip, End Fitting, with Retainer Nut, Ace Glass Incorporated
Supplier: Ace Glass
5857-05 5857-10 5857-15 5857-20
89052-994EA 484.98 CAD
89052-994 89052-996 89052-998 89053-000
Adapter, Bottom Drip, End Fitting, with Retainer Nut, Ace Glass Incorporated
Fittings and Connectors
These adapters are for use as column end fittings that allow connection of appropriate size Ace-Thred to tubing connectors with NPT threads.
  • Locking retainer nut
  • Micro screen and o-ring included
  • Machined of polypropylene
Adapters feature a "coin" tightened locking retainer nut that holds a 350 micron support screen firmly in place.
Ordering information: Each adapter is supplied with a retainer nut lock (89053-034 for #25; 89053-036 for #50), 350 micro screen (89052-200 for #25; 89052-202), and FETFE o-ring (89061-004 for #25; 89061-008 for #50).