Chromatography Column, Ace Glass Incorporated
Supplier: Ace Glass
5820-04 5820-08 5820-104 5820-12 5820-16 5820-20 5820-24 5820-30 5820-34 5820-37 5820-40 5820-50 5820-53 5820-55 5820-58 5820-59
89052-334EA 126.67 CAD
89052-334 89052-338 89052-340 89052-352 89052-362 89052-366 89052-370 89052-372 89052-374 89052-378 89052-382 89052-384 89052-386 89052-388 89052-392 89052-394
Chromatography Column, Ace Glass Incorporated
Chromatography Columns
Rugged chromatography column with internally threaded ends to which a wide choice of upper and lower end pieces can be fitted.
Available in the diameters and lengths listed below. Fabricated from borosilicate glass.